Learn More
Novus Biologicals™ Human 20S Proteasome ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP262173
Description
The Human 20S Proteasome ELISA Kit (Colorimetric) measures human 20S Proteasome in biological samples.Specifications
25 - 1600 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
1 Kit | |
20S Proteasome |
25 ng/mL | |
Biological Sample | |
Human | |
Store at -20°C. Avoid freeze-thaw cycles. |