Learn More
Gibco™ Human 4-1BB Ligand (TNFSF9), Animal-Free Recombinant Protein, PeproTech®

E. coli Recombinant Protein
$241.00 - $3564.00
Specifications
Accession Number | P41273 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 8744 |
Molecular Weight (g/mol) | 19.5 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AF31011100
![]() ![]() |
Gibco™
AF31011100UG |
100 µg |
Each for $740.00
|
|
|||||
AF310111MG
![]() ![]() |
Gibco™
AF310111MG |
1 mg |
Each for $3,564.00
|
|
|||||
AF3101120UG
![]() ![]() |
Gibco™
AF3101120UG |
20 µg |
Each for $241.00
|
|
|||||
AF31011500
![]() ![]() |
Gibco™
AF31011500UG |
500 µg |
Each for $2,139.00
|
|
|||||
Description
CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.Specifications
P41273 | |
protein with no preservative | |
19.5 | |
E. coli | |
RUO | |
4 1 BB Ligand; 4 1 BBL; 41 BB Ligand; 4-1 BB ligand; 41 BBL; 4-1 BBL; 4-1 BB-L; AI848817; CD 137 Ligand; CD 137 L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1 BB-L; Ly63l; receptor 4-1 BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5 A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
8744 | |
Human 4-1 BB Ligand (TNFSF9), Animal-Free | |
-20°C | |
<0.1 EU/µg | |
TNFSF9 | |
Unconjugated | |
MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.