Learn More
Novus Biologicals Human 5-Lipoxygenase ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP266367
Description
The Human 5-Lipoxygenase ELISA Kit (Colorimetric) measures human 5-Lipoxygenase in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent. | |
Human | |
1 Kit |
This kit recognizes Human 5-LO in samples. No significant cross-reactivity or interference between Human 5-LO and analogues was observed. | |
240 | |
0.10 ng/mL | |
5-Lipoxygenase | |
ALOX5 | |
Biological Fluids, Plasma, Serum |