Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human A2ML1 Full-length ORF (AAH93840.1, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00144568P01S

 View more versions of this product

Catalog No. 89-932-032


Add to cart

Description

Description

The alpha-macroglobulin (AM) superfamily of proteins contains both complement components and protease inhibitors, including A2M (MIM 103950) and A2ML1. AM proteins display a unique trap mechanism of inhibition, by which the AM inhibitor undergoes a major conformational change upon its cleavage by a protease, thus trapping the protease and blocking it from subsequent substrate binding (Galliano et al., 2006 [PubMed 16298998]).[supplied by OMIM]

Sequence: MYTLEASGQGCVYVQTVLRYNILPPTNMKTFSLSVEIGKARCEQPTSPRSLTLTIHTSYVGSRSSSNMAIVEVKMLSGFSPMEGTNQLLLQQPLVKKVEFGTDTLNIYLDELIKNTQTYTFTISQSVLVTNLKPATIKVYDYYLPDEQATIQYSDPCE
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
A2ML1
Wheat Germ (in vitro)
GST
AAH93840.1
Liquid
144568
A2ML1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CPAMD9/DKFZp686C1729/DKFZp686D2011/DKFZp686G1812/DKFZp686L1821/DKFZp686O1010/FLJ16045/FLJ25179/FLJ39129/FLJ41597/FLJ41598/FLJ41607
A2ML1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit