Learn More
Abnova™ Human A2ML1 Full-length ORF (AAH93840.1, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00144568P01L
Description
The alpha-macroglobulin (AM) superfamily of proteins contains both complement components and protease inhibitors, including A2M (MIM 103950) and A2ML1. AM proteins display a unique trap mechanism of inhibition, by which the AM inhibitor undergoes a major conformational change upon its cleavage by a protease, thus trapping the protease and blocking it from subsequent substrate binding (Galliano et al., 2006 [PubMed 16298998]).[supplied by OMIM]
Sequence: MYTLEASGQGCVYVQTVLRYNILPPTNMKTFSLSVEIGKARCEQPTSPRSLTLTIHTSYVGSRSSSNMAIVEVKMLSGFSPMEGTNQLLLQQPLVKKVEFGTDTLNIYLDELIKNTQTYTFTISQSVLVTNLKPATIKVYDYYLPDEQATIQYSDPCESpecifications
AAH93840.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
A2ML1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
144568 | |
A2ML1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPAMD9/DKFZp686C1729/DKFZp686D2011/DKFZp686G1812/DKFZp686L1821/DKFZp686O1010/FLJ16045/FLJ25179/FLJ39129/FLJ41597/FLJ41598/FLJ41607 | |
A2ML1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.