Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human A4GALT Full-length ORF (NP_059132.1, 1 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00053947P01

Catalog No. 89-932-037


Add to cart

Description

Description

The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor. [provided by RefSeq]

Sequence: MSKPPDLLLRLLRGAPRQRVCTLFIIGFKFTFFVSIMIYWHVVGEPKEKGQLYNLPAEIPCPTLTPPTPPSHGPTPGNIFFLETSDRTNPNFLFMCSVESAARTHPESHVLVLMKGLPGGNASLPRHLGISLLSCFPNVQMLPLDLRELFRDTPLADWYAAVQGRWEPYLLPVLSDASRIALMWKFGGIYLDTDFIVLKNLRNLTNVLGTQSRYVLNGAFLAFERRHEFMALCMRDFVDHYNGWIWGHQGPQLLTRVFKKWCSIRSLAESRACRGVTTLPPEAFYPIPWQDWKKYFEDINPEELPRLLSATYAVHVWNKKSQGTRFEATSRALLAQLHARYCPTTHEAMKMYL
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.9kDa
Glutathione Sepharose 4 Fast Flow
2 ug
RUO
A4GALT
Wheat Germ (in vitro)
GST
NP_059132.1
Liquid
53947
A4GALT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A14GALT/A4GALT1/P1/PK
A4GALT
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit