Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABCB10 Partial ORF (NP_036221, 233 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023456Q01S

 View more versions of this product

Catalog No. 89-932-107


Add to cart

Description

Description

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown. [provided by RefSeq]

Sequence: VYLMQTSGQRIVNRLRTSLFSSILRQEVAFFDKTRTGELINRLSSDTALLGRSVTENLSDGLRAGAQASVGISMMFFVS
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.43kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VYLMQTSGQRIVNRLRTSLFSSILRQEVAFFDKTRTGELINRLSSDTALLGRSVTENLSDGLRAGAQASVGISMMFFVS
RUO
ABCB10
Wheat Germ (in vitro)
GST
NP_036221
Liquid
23456
ABCB10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EST20237/M-ABC2/MTABC2
ABCB10
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit