Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ABCB5 Full-length ORF (ADR83259.1) Recombinant Protein without tag
Used for AP, Func, Screening
Supplier: Abnova™ H00340273G01
Description
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules (Chen et al., 2005 [PubMed 15760339]).[supplied by OMIM]
Sequence: MVDENDIRALNVRHYRDHIGVVSQEPVLFGTTISNNIKYGRDDVTDEEMERAAREANAYDFIMEFPNKFNTLVGEKGAQMSGGQKQRIAIARALVRNPKILILDEATSALDSESKSAVQAALEKASKGRTTIVVAHRLSTIRSADLIVTLKDGMLAEKGAHAELMAKRGLYYSLVMSQDIKKADEQMESMTYSTERKTNSLPLHSVKSIKSDFIDKAEESTQSKEISLPEVSLLKILKLNKPEWPFVVLGTLASVLNGTVHPVFSIIFAKIITMFGNNDKTTLKHDAEIYSMIFVILGVICFVSYFMQGLFYGRAGEILTMRLRHLAFKAMLYQDIAWFDEKENSTGGLTTILAIDIAQIQGATGSRIGVLTQNATNMGLSVIISFIYGWEMTFLILSIAPVLAVTGMIETAAMTGFANKDKQELKHAGKIATEALENIRTIVSLTREKAFEQMYEEMLQTQHRNTSKKAQIIGSCYAFSHAFIYFAYAAGFRFGAYLIQAGRMTPEGMFIVFTAIAYGAMAIGETLVLAPEYSKAKSGAAHLFALLEKKPNIDSRSQEGKKPDTCEGNLEFREVSFFYPCRPDVFILRGLSLSIERGKTVAFVGSSGCGKSTSVQLLQRLYDPVQGQVLFDGVDAKELNVQWLRSQIAIVPQEPVLFNCSIAENIAYGDNSRVVPLDEIKEAANAANIHSFIEGLPEKYNTQVGLKGAQLSGGQKQRLAIARALLQKPKILLLDEATSALDNDSEKVVQHALDKARTGRTCLVVTHRLSAIQNADLIVVLHNGKIKEQGTHQELLRNRDIYFKLVNAQSVQSpecifications
ADR83259.1 | |
25mM Tris-HCl of pH8.0 containing 2% glycerol. | |
89.4kDa | |
2 ug | |
RUO | |
ABCB5 | |
Wheat Germ (in vitro) | |
None | |
Liquid |
Antibody Production, Compound Screening, Functional Study | |
340273 | |
ABCB5 (Human) Recombinant Protein | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABCB5alpha/ABCB5beta/EST422562 | |
ABCB5 | |
Recombinant | |
wheat germ expression system with proprietary liposome technology |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction