Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABCC13 Full-length ORF (ENSP00000345983, 1 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00150000P01L

 View more versions of this product

Catalog No. 89-932-128


Add to Cart

Description

Description

This gene is a member of the superfamily of genes encoding ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This family member is part of the MRP subfamily, which is involved in multi-drug resistance, but the human locus is now thought to be a pseudogene incapable of encoding a functional ABC protein. Alternative splicing results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq]

Sequence: MLSSTQNAGGSYQRVRGALDTQKCSPEKSASFFSKVTYSWFSRVITLGYKRPLEREDLFELKESDSFCTACPIFEKQWRKEVLRNQERQKVKVSCYKEAHIKKPSLLYALWNTFKSILIQVALFKVFADILSFTSPLIMNYTRKVNYLMGLPCENQKITSYSQASGRDS
Specifications

Specifications

ENSP00000345983
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ABCC13
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
150000
ABCC13 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C21orf73/PRED6
ABCC13
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit