Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABCC6 Partial ORF (NP_001162, 831 a.a. - 930 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000368Q01L

 View more versions of this product

Catalog No. 89-932-138


Add to cart

Description

Description

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq]

Sequence: IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRV
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRV
RUO
ABCC6
Wheat Germ (in vitro)
GST
NP_001162
Liquid
368
ABCC6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABC34/ARA/EST349056/MLP1/MOATE/MRP6/PXE/PXE1
ABCC6
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit