Learn More
Abnova™ Human ABHD5 Full-length ORF (AAH21958, 1 a.a. - 349 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051099P01S
Description
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation. [provided by RefSeq]
Sequence: MAAEEEEVDSADTGERSGWLTGWLPTWCPTSISHLKEAEEKMLKCVPCTYKKEPVRISNGNKIWTLKFSHNISNKTPLVLLHGFGGGLGLWALNFGDLCTNRPVYAFDLLGFGRSSRPRFDSDAEEVENQFVESIEEWRCALGLDKMILLGHNLGGFLAAAYSLKYPSRVNHLILVEPWGFPERPDLADQDRPIPVWIRALGAALTPFNPLAGLRIAGPFGLSLVQRLRPDFKRKYSSMFEDDTVTEYIYHCNVQTPSGETAFKNMTIPYGWAKRPMLQRIGKMHPDIPVSVIFGARSCIDGNSGTSIQSLRPHSYVKTIAILGAGHYVYADQPEEFNQKVKEICDTVDSpecifications
AAH21958 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.13kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ABHD5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51099 | |
ABHD5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDS/CGI58/IECN2/MGC8731/NCIE2 | |
ABHD5 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.