Learn More
Abnova™ Human ABL2 (NM_005158, 38 a.a. - end) Partial Recombinant with His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3834
Catalog No. 89-932-226
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
Insect cell (Sf9) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
27
ABL2 (Human) Recombinant Protein
>70% by densitometry
5 ug
RUO
ABL2
The specific activity was determined to be 1218nmol /min/mg as per Activity Assay Protocol.
Yes
NM_005158
In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
132kDa
Insect cell (Sf9) expression system
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABLL/ARG/FLJ22224/FLJ31718/FLJ41441
ABL2
Insect
His