Learn More
Abnova™ Human ABT1 Full-length ORF (NP_037507.1, 1 a.a. - 272 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_037507.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 29777 |
Molecular Weight (g/mol) | 57.5kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-932-249
|
Abnova™
H00029777P02L |
25 ug |
Each for $769.00
|
|
|||||
89-932-248
|
Abnova™
H00029777P02S |
10 ug |
Each for $499.00
|
|
|||||
Description
Basal transcription of genes by RNA polymerase II requires the interaction of TATA-binding protein (TBP) with the core region of class II promoters. Studies in mouse suggest that the protein encoded by this gene likely activates basal transcription from class II promoters by interaction with TBP and the class II promoter DNA. [provided by RefSeq]
Sequence: MEAEESEKAATEQEPLEGTEQTLDAEEEQEESEEAACGSKKRVVPGIVYLGHIPPRFRPLHVRNLLSAYGEVGRVFFQAEDRFVRRKKKAAAAAGGKKRSYTKDYTEGWVEFRDKRIAKRVAASLHNTPMGARRRSPFRYDLWNLKYLHRFTWSHLSEHLAFERQVRRQRLRAEVAQAKRETDFYLQSVERGQRFLAADGDPARPDGSWTFAQRPTEQELRARKAARPGGRERARLATAQDKARSNKGLLARIFGAPPPSESMEGPSLVRDSSpecifications
NP_037507.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
hABT1 | |
ABT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
29777 | |
ABT1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ABT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.