Learn More
Novus Biologicals™ Human ACD ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266386
Catalog No. NBP266386
Add to cart
Description
Description
The Human ACD ELISA Kit (Colorimetric) measures human ACD in serum, plasma and other biological fluids.
Specifications
Specifications
ELISA Kit (Colorimetric)
This kit recognizes Human ACD in samples. No significant cross-reactivity or interference between Human ACD and analogues was observed.
65057
ACD
1 Kit
18.75 pg/mL
31.25 - 2000 pg/mL
ELISA
Storage is content dependent.
Human
Biological Fluids, Plasma, Serum
ACD