Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ACE-2 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems FAB933S100UG
This item is not returnable.
View return policy
Description
ACE-2 Polyclonal specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry.Specifications
ACE-2 | |
Polyclonal | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BYF1 | |
Goat | |
Antigen Affinity-purified | |
RUO | |
59272 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
Mouse myeloma cell line NS0-derived recombinant human ACE-2, Gln18-Ser740, Accession # Q9BYF1 | |
100 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction