Learn More
Novus Biologicals™ Human Acetyl-CoA Carboxylase alpha/ACACA ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266384
Catalog No. NBP266384
Add to Cart
Description
Description
The Human Acetyl-CoA Carboxylase alpha/ACACA ELISA Kit (Colorimetric) measures human Acetyl-CoA Carboxylase alpha/ACACA in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 - 100 ng/mL
This kit recognizes Human ACC in samples. No significant cross-reactivity or interference between Human ACC and analogues was observed.
Biological Fluids, Plasma, Serum
31
Human
Storage of components varies. See protocol for specific instructions.
0.94 ng/mL
ELISA Kit (Colorimetric)
ELISA
ACACA
1 Kit
Acetyl-CoA Carboxylase alpha/ACACA
Product Suggestions
Product Suggestions