Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACO1 Full-length ORF (NP_002188.1, 1 a.a. - 889 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000048P02S

 View more versions of this product

Catalog No. 89-932-339


Add to cart

Description

Description

Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes. [provided by RefSeq]

Sequence: MSNPFAHLAEPLDPVQPGKKFFNLNKLEDSRYGRLPFSIRVLLEAAIRNCDEFLVKKQDIENILHWNVTQHKNIEVPFKPARVILQDFTGVPAVVDFAAMRDAVKKLGGDPEKINPVCPADLVIDHSIQVDFNRRADSLQKNQDLEFERNRERFEFLKWGSQAFHNMRIIPPGSGIIHQVNLEYLARVVFDQDGYYYPDSLVGTDSHTTMIDGLGILGWGVGGIEAEAVMLGQPISMVLPQVIGYRLMGKPHPLVTSTDIVLTITKHLRQVGVVGKFVEFFGPGVAQLSIADRATIANMCPEYGATAAFFPVDEVSITYLVQTGRDEEKLKYIKKYLQAVGMFRDFNDPSQDPDFTQVVELDLKTVVPCCSGPKRPQDKVAVSDMKKDFESCLGAKQGFKGFQVAPEHHNDHKTFIYDNTEFTLAHGSVVIAAITSCTNTSNPSVMLGAGLLAKKAVDAGLNVMPYIKTSLSPGSGVVTYYLQESGVMPYLSQLGFDVVGYGCMTCIGNSGPLPEPVVEAITQGDLVAVGVLSGNRNFEGRVHPNTRANYLASPPLVIAYAIAGTIRIDFEKEPLGVNAKGQQVFLKDIWPTRDEIQAVERQYVIPGMFKEVYQKIETVNESWNALATPSDKLFFWNSKSTYIKSPPFFENLTLDLQPPKSIVDAYVLLNLGDSVTTDHISPAGNIARNSPAARYLTNRGLTPREFNSYGSRRGNDAVMARGTFANIRLLNRFLNKQAPQTIHLPSGEILDVFDAAERYQQAGLPLIVLAGKEYGAGSSRDWAAKGPFLLGIKAVLAESYERIHRSNLVGMGVIPLEYLPGENADALGLTGQERYTIIIPENLKPQMKVQVKLDTGKTFQAVMRFDTDVELTYFLNGGILNYMIRKMAK
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
124.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ACO1
Wheat Germ (in vitro)
GST
NP_002188.1
Liquid
48
ACO1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACONS/IREB1/IREBP/IREBP1/IRP1
ACO1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit