Learn More
Novus Biologicals™ Human Aconitase 2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266394
Catalog No. NBP266394
Add to cart
Description
Description
The Human Aconitase 2 ELISA Kit (Colorimetric) measures human Aconitase 2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Human ACO-2 in samples. No significant cross-reactivity or interference between human ACO-2 and analogues was observed.
50
Human
1 Kit
Storage is content dependent.
0.19 ng/mL
ELISA
ACO2
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
Aconitase 2