Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACOX2 Full-length ORF (NP_003491.1, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008309P01S

 View more versions of this product

Catalog No. 89-932-352


Add to cart

Description

Description

The product of this gene belongs to the acyl-CoA oxidase family. It encodes the branched-chain acyl-CoA oxidase which is involved in the degradation of long branched fatty acids and bile acid intermediates in peroxisomes. Deficiency of this enzyme results in the accumulation of branched fatty acids and bile acid intermediates, and may lead to Zellweger syndrome, severe mental retardation, and death in children. [provided by RefSeq]

Sequence: MGSPVHRVSLGDTWSRQMHPDIESERYMQSFDVERLTNILDGGAQNTALRRKVESIIHSYPEFSCKDNYFMTQNERYKAAMRRAFHIRLIARRLGWLEDGRELGYAYRALSGDVALNIHRVFVRALRSLGSEEQIAKWDPLCKNIQIIATYAQTELGHGTYLQGLETEATYDAATQEFVIHSPTLTATKWWPGDLGRSATHALVQAQLICSGARRGMHAFIVPIRSLQDHTPLPGIIIGDIGPKMDFDQTDNGFLQLNHVRVPRENMLSRFAQVLPDGTYVKLGTAQSNYLPMVVVRVELLSGEILPILQKACVIAMRYSVIRRQSRLRPSDPEAKVLDYQTQQQKLFPQLAISYAFHFLAVSLLEFFQHSYTAILNQDFSFLPELHALSTGMKAMMSEFCTQGAEMCRRACGGHGYSKLSGLPSLVTKLSASCTYEGENTVLYLQVARFLVKSYLQTQMSPGSTPQRSLSPSVAYLTAPDLARCPAQRAADFLCPELYTTAWAHVAVRLIKDSVQHLQTLTQSGADQHEAWNQTTVIHLQAAKVHCYYVTVKGFTEALEKLENEPAIQQVLKRLCDLHAIHGILTNSGDFLHDAFLSGAQVDMARTAYLDLLRLIRKDAILLTDAFDFTDQCLNSALGCYDGNVYERLFQWAQKSPTNTQENPAYEEYIRPLLQSWRSKL
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
103.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ACOX2
Wheat Germ (in vitro)
GST
NP_003491.1
Liquid
8309
ACOX2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCOX/BRCACOX/BRCOX/THCCox
ACOX2
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit