Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACOX3 Full-length ORF (AAH17053.1, 1 a.a. - 624 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name ACOX3 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 96kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-932-359
View Documents
Abnova™
H00008310P01S
10 ug
Each for $460.50
Add to cart
 
89-932-360
View Documents
Abnova™
H00008310P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

Acyl-Coenzyme A oxidase 3 also know as pristanoyl -CoA oxidase (ACOX3)is involved in the desaturation of 2-methyl branched fatty acids in peroxisomes. Unlike the rat homolog, the human gene is expressed in very low amounts in liver such that its mRNA was undetectable by routine Northern-blot analysis or its product by immunoblotting or by enzyme activity measurements. However the human cDNA encoding a 700 amino acid protein with a peroxisomal targeting C-terminal tripeptide S-K-L was isolated and is thought to be expressed under special conditions such as specific developmental stages or in a tissue specific manner in tissues that have not yet been examined. [provided by RefSeq]

Sequence: MASTVEGGDTALLPEFPRGPLDAYRARASFSWKELALFTEGEGMLRFKKTIFSALENDPLFARSPGADLSLEKYRELNFLRCKRIFEYDFLSVEDMFKSPLKVPALIQCLGMYDSSLAAKYLLHSLVFGSAVYSSGSERHLTYIQKIFRMEIFGCFALTELSHGSNTKAIRTTAHYDPATEEFIIHSPDFEAAKFWVGNMGKTATHAVVFAKLCVPGDQCHGLHPFIVQIRDPKTLLPMPGVMVGDIGKKLGQNGLDNGFAMFHKVRVPRQSLLNRMGDVTPEGTYVSPFKDVRQRFGASLGSLSSGRVSIVSLAILNLKLAVAIALRFSATRRQFGPTEEEEIPVLEYPMQQWRLLPYLAAVYALDHFSKSLFLDLVELQRGLASGDRSARQAELGREIHALASASKPLASWTTQQGIQECREACGGHGYLAMNRLGVLRDDNDPNCTYEGDNNILLQQTSNYLLGLLAHQVHDGACFRSPLKSVDFLDAYPGILDQKFEVSSVADCLDSAVALAAYKWLVCYLLRETYQKLNQEKRSGSSDFEARNKCQVSHGRPLALAFVELTVVQRFHEHVHQPSVPPSLRAVLGRLSALYALWSLSRHAALLYRAERRCSCPGRRDRSS
Specifications

Specifications

ACOX3 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
96kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit