Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACPT Full-length ORF (AAI46507.1, 1 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00093650P01L

 View more versions of this product

Catalog No. 89-932-379


Add to cart

Description

Description

Acid phosphatases are enzymes capable of hydrolyzing orthophosphoric acid esters in an acid medium. This gene is up-regulated by androgens and is down-regulated by estrogens in the prostate cancer cell line. This gene exhibits a lower level of expression in testicular cancer tissues than in normal tissues. The protein encoded by this gene has structural similarity to prostatic and lysosomal acid phosphatases. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: MAGLGFWGHPAGPLLLLLLLVLPPRALPEGPLVFVALVFRHGDRAPLASYPMDPHKEVASTLWPRGLGQLTTEGVRQQLELGRFLRSRYEAFLSPEYRREEVYIRSTDFDRTLESAQANLAGLFPEAAPGSPEARWRPIPVHTVPVAEDKLLRFPMRSCPRYHELLREATEAAEYQEALEGWTGFLSRLENFTGLSLVGEPLRRAWKVLDTLMCQQAHGLPLPAWASPDVLRTLAQISALDIGAHVGPPRAAEKAQLTGGILLNAILANFSRVQRLGLPLKMVMYSAHDSTLLALQGALGLYDGHTPPYAACLGFEFRKHLGNPAKDGGNVTVSLFYRNDSAHLPLPLSLPGCPAPCPLGRFYQLTAPARPPAHGVSCHGPYEAAIPPAPVVPLLAGAVAVLVALSLGLGLLAWRPGCLRALGGPV
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.81kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ACPT
Yes
AAI46507.1
Liquid
93650
ACPT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACPT
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit