Learn More
Abnova™ Human ACRBP Full-length ORF (NP_115878.2, 1 a.a. - 543 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00084519P01L
Description
The protein encoded by this gene is similar to proacrosin binding protein sp32 precursor found in mouse, guinea pig, and pig. This protein is located in the sperm acrosome and is thought to function as a binding protein to proacrosin for packaging and condensation of the acrosin zymogen in the acrosomal matrix. This protein is a member of the cancer/testis family of antigens and it is found to be immunogenic. In normal tissues, this mRNA is expressed only in testis, whereas it is detected in a range of different tumor types such as bladder, breast, lung, liver, and colon. [provided by RefSeq]
Sequence: MRKPAAGFLPSLLKVLLLPLAPAAAQDSTQASTPGSPLSPTEYERFFALLTPTWKAETTCRLRATHGCRNPTLVQLDQYENHGLVPDGAVCSNLPYASWFESFCQFTHYRCSNHVYYAKRVLCSQPVSILSPNTLKEIEASAEVSPTTMTSPISPHFTVTERQTFQPWPERLSNNVEELLQSSLSLGGQEQAPEHKQEQGVEHRQEPTQEHKQEEGQKQEEQEEEQEEEGKQEEGQGTKEGREAVSQLQTDSEPKFHSESLSSNPSSFAPRVREVESTPMIMENIQELIRSAQEIDEMNEIYDENSYWRNQNPGSLLQLPHTEALLVLCYSIVENTCIITPTAKAWKYMEEEILGFGKSVCDSLGRRHMSTCALCDFCSLKLEQCHSEASLQRQQCDTSHKTPFVSPLLASQSLSIGNQVGSPESGRFYGLDLYGGLHMDFWCARLATKGCEDVRVSGWLQTEFLSFQDGDFPTKICDTDYIQYPNYCSFKSQQCLMRNRNRKVSRMRCLQNETYSALSPGKSEDVVLRWSQEFSTLTLGQFGSpecifications
NP_115878.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ACRBP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84519 | |
ACRBP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ51160/OY-TES-1/SP32 | |
ACRBP | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.