Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ACSS2 Partial ORF (NP_061147.1, 32 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055902Q01L

 View more versions of this product

Catalog No. 89-932-415


Add to cart

Description

Description

This gene encodes a cytosolic enzyme that catalyzes the activation of acetate for use in lipid synthesis and energy generation. The protein acts as a monomer and produces acetyl-CoA from acetate in a reaction that requires ATP. Expression of this gene is regulated by sterol regulatory element-binding proteins, transcription factors that activate genes required for the synthesis of cholesterol and unsaturated fatty acids. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: PPEVSRSAHVPSLQRYRELHRRSVEEPREFWGDIAKEFYWKTPCPGPFLRYNFDVTKGKIFIEWMKGATTNICYNVLDRNVHEKKLGDKVAFYWEGNEP
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PPEVSRSAHVPSLQRYRELHRRSVEEPREFWGDIAKEFYWKTPCPGPFLRYNFDVTKGKIFIEWMKGATTNICYNVLDRNVHEKKLGDKVAFYWEGNEP
RUO
ACSS2
Wheat Germ (in vitro)
GST
NP_061147.1
Liquid
55902
ACSS2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACAS2/ACS/ACSA/AceCS/DKFZp762G026/dJ1161H23.1
ACSS2
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit