Learn More
Novus Biologicals™ Human Activin B/Inhibin beta B ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266432
Catalog No. NBP266432
Add to Cart
Description
Description
The Human Activin B/Inhibin beta B ELISA Kit (Colorimetric) measures human Activin B/Inhibin beta B in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
This kit recognizes Human ACVB in samples. No significant cross-reactivity or interference between Human ACVB and analogues was observed.
Biological Fluids, Plasma, Serum
3625
Human
Storage of components varies. See protocol for specific instructions.
9.38 pg/mL
ELISA Kit (Colorimetric)
ELISA
INHBB
1 Kit
Activin B/Inhibin beta B
Recommendations
Recommendations