Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin C/Inhibin beta C Antibody, Unconjugated, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Activin C/Inhibin beta C |
---|---|
Concentration | LYOPH |
Dilution | ELISA |
Applications | ELISA, Immunohistochemistry |
Classification | Polyclonal |
Description
Activin C/Inhibin beta C Polyclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Activin C/Inhibin beta C | |
ELISA | |
Polyclonal | |
Goat | |
Human | |
inhibin beta C chain, IHBC, INHBC, inhibin, beta C | |
INHBC | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
ELISA, Immunohistochemistry | |
Unconjugated | |
RUO | |
3626 | |
3626 | |
E. coli-derived recombinant human Activin C Gly237-Ser352 Accession # 3626 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Activin C beta C subunit in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title