Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ACVR1B (aa 32-126) His Tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

CHO cells Recombinant Protein

$96.30 - $1798.00

Specifications

Recombinant Yes
Name Human ACVR1B (aa 32-126) His Tag
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87813 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87813
10 µg
Each for $96.30
Add to cart
 
PIRP87814 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87814
25 µg
Each for $173.40
Add to cart
 
PIRP87815 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87815
100 µg
Each for $449.50
Add to cart
 
PIRP87816 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87816
500 µg
Each for $1,798.00
Add to cart
 
Description

Description

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I (I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with a cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling, and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. This gene encodes activin A type IB receptor, composed of 11 exons. Alternative splicing and alternative polyadenylation result in 3 fully described transcript variants. The mRNA expression of variants 1, 2, and 3 is confirmed, and a potential fourth variant contains an alternative exon 8 and lacks exons 9 through 11, but its mRNA expression has not been confirmed.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative
CHO cells
-20°C
P36896
ACVR1B
>95%
Human ACVR1B (aa 32-126) His Tag
200 µg/mL
Liquid
6820432J04; activin A receptor type 1 B; activin A receptor type IB; activin A receptor, type 1 B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1 B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; ACVR1B; ACVRLK2; Acvrlk4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI
RUO
Human
91
38.3 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit