Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAM30 Partial ORF (NP_068566, 199 a.a. - 298 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name ADAM30 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.74kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-932-561
View Documents
Abnova™
H00011085Q01S
10 ug
Each for $460.50
Add to cart
 
89-932-562
View Documents
Abnova™
H00011085Q01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats. [provided by RefSeq]

Sequence: YKHPKYLELILLFDQSRYRFVNNNLSQVIHDAILLTGIMDTYFQDVRMRIHLKALEVWTDFNKIRVGYPELAEVLGRFVIYKKSVLNARLSSDWAHLYLQ
Specifications

Specifications

ADAM30 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.74kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit