Learn More
Abnova™ Human ADAMTS1 (253 a.a. - 617 a.a.) amino acids 618-968 deleted Partial Recombinant Protein with His tag expressed in insect cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P4930
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two disintegrin loops and three C-terminal TS motifs and has anti-angiogenic activity. The expression of this gene may be associated with various inflammatory processes as well as development of cancer cachexia. This gene is likely to be necessary for normal growth, fertility, and organ morphology and function. [provided by RefSeq]
Sequence: FVSSHRYVETMLVADQSMAEFHGSGLKHYLLTLFSVAARLYKHPSIRNSVSLVVVKILVIHDEQKGPEVTSNAALTLRNFCNWQKQHNPPSDRDAEHYDTAILFTRQDLCGSQTCDTLGMADVGTVCDPSRSCSVIEDDGLQAAFTTAHELGHVFNMPHDDAKQCASLNGVNQDSHMMASMLSNLDHSQPWSPCSAYMITSFLDNGHGECLMDKPHNPIQLPGDLPGTSYDANRQCQFTFGEDSKHCPDAASTCSTLWCTGTSGGVLVCQTKHFPWADGTSCGEGKWCINGKCVNKTDRKHFDTPFHGNWGMWGPWGDCSRTCGGGVQYTMRECDNPVPKNGGKYCEGKRVRYRSCNLEDCPDNSpecifications
Q9UHI8 | |
Functional Study, SDS-PAGE | |
9510 | |
ADAMTS1 (d618-968) (Human) Recombinant Protein | |
15% SDS-PAGE Stained with Coomassie Blue | |
FVSSHRYVETMLVADQSMAEFHGSGLKHYLLTLFSVAARLYKHPSIRNSVSLVVVKILVIHDEQKGPEVTSNAALTLRNFCNWQKQHNPPSDRDAEHYDTAILFTRQDLCGSQTCDTLGMADVGTVCDPSRSCSVIEDDGLQAAFTTAHELGHVFNMPHDDAKQCASLNGVNQDSHMMASMLSNLDHSQPWSPCSAYMITSFLDNGHGECLMDKPHNPIQLPGDLPGTSYDANRQCQFTFGEDSKHCPDAASTCSTLWCTGTSGGVLVCQTKHFPWADGTSCGEGKWCINGKCVNKTDRKHFDTPFHGNWGMWGPWGDCSRTCGGGVQYTMRECDNPVPKNGGKYCEGKRVRYRSCNLEDCPDN | |
RUO | |
ADAMTS1 | |
≥ 1.4mU/mg | |
Recombinant | |
Insect cell expression system | |
>90% by SDS-PAGE |
0.2 mg/mL | |
Liquid | |
41kDa | |
Insect cell expression system | |
5 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3-C5/KIAA1346/METH1 | |
ADAMTS1 | |
Baculovirus | |
His | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.