Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAMTS17 Partial ORF (NP_620688, 543 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00170691Q01L

 View more versions of this product

Catalog No. 89-932-589


Add to cart

Description

Description

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by ADAMTS19, another family member. The function of this protein has not been determined. [provided by RefSeq]

Sequence: DGDWSPWGAWSMCSRTCGTGARFRQRKCDNPPPGPGGTHCPGASVEHAVCENLPCPKGLPSFRDQQCQAHDRLSPKKKGLLTAVVVDDKPCELYCSPLGKESPLLVAD
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DGDWSPWGAWSMCSRTCGTGARFRQRKCDNPPPGPGGTHCPGASVEHAVCENLPCPKGLPSFRDQQCQAHDRLSPKKKGLLTAVVVDDKPCELYCSPLGKESPLLVAD
RUO
ADAMTS17
Wheat Germ (in vitro)
GST
NP_620688
Liquid
170691
ADAMTS17 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16363/FLJ32769
ADAMTS17
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit