Learn More
Abnova™ Human ADAMTS5 amino acids 625-930 deleted Recombinant Protein with His tag expressed in insect cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4931
Catalog No. 89-932-603
Add to cart
Description
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs and functions as aggrecanase to cleave aggrecan, a major proteoglycan of cartilage. [provided by RefSeq]
Specifications
Specifications
Insect cell expression system
0.1 mg/mL
Liquid
11096
ADAMTS5 (d625-930) (Human) Recombinant Protein
≥50% by SDS-PAGE
5 ug
RUO
ADAMTS5
≥ 5mU/mg
Yes
Q9UNA0
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
40.8kDa
Insect cell expression system
15% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADAMTS11/ADMP-2/FLJ36738
ADAMTS5
Insect
His