Learn More
Novus Biologicals™ Human ADAMTS5 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266444
Catalog No. NBP266444
Add to Cart
Description
Description
The Human ADAMTS5 ELISA Kit (Colorimetric) measures human ADAMTS5 in serum, plasma and other biological fluids.
Specifications
Specifications
0.78 - 50 ng/mL
This kit recognizes Human ADAMTS5 in samples. No significant cross-reactivity or interference between Human ADAMTS5 and analogues was observed.
Biological Fluids, Plasma, Serum
11096
Human
Storage of components varies. See protocol for specific instructions.
0.47 ng/mL
ELISA Kit (Colorimetric)
ELISA
ADAMTS5
1 Kit
ADAMTS5
Recommendations
Recommendations