Learn More
Novus Biologicals™ Human ADCY4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP266643
Description
The Human ADCY4 ELISA Kit (Colorimetric) measures human ADCY4 in serum, plasma and other biological fluids.Specifications
0.63 - 40 ng/mL | |
This kit recognizes Human ADCY4 in samples. No significant cross-reactivity or interference between Human ADCY4 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
196883 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
0.38 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
ADCY4 | |
1 Kit | |
ADCY4 |