Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADCY8 Partial ORF (NP_001106, 1142 a.a. - 1251 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000114Q01L

 View more versions of this product

Catalog No. 89-932-653


Add to cart

Description

Description

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase [provided by RefSeq]

Sequence: DQGFAFDYRGEIYVKGISEQEGKIKTYFLLGRVQPNPFILPPRRLPGQYSLAAVVLGLVQSLNRQRQKQLLNENNNTGIIKGHYNRRTLLSPSGTEPGAQAEGTDKSDLP
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DQGFAFDYRGEIYVKGISEQEGKIKTYFLLGRVQPNPFILPPRRLPGQYSLAAVVLGLVQSLNRQRQKQLLNENNNTGIIKGHYNRRTLLSPSGTEPGAQAEGTDKSDLP
RUO
ADCY8
Wheat Germ (in vitro)
GST
NP_001106
Liquid
114
ADCY8 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AC8/ADCY3/HBAC1
ADCY8
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit