Learn More
Novus Biologicals™ Human Adenosine A2aR ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266650
Catalog No. NBP266650
Add to Cart
Description
Description
The Human Adenosine A2aR ELISA Kit (Colorimetric) measures human Adenosine A2aR in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Human ADORA2a in samples. No significant cross-reactivity or interference between Human ADORA2a and analogues was observed.
Biological Fluids, Plasma, Serum
135
Human
GPCR
Adenosine A2a R
0.19 ng/mL
ELISA Kit (Colorimetric)
ELISA
ADORA2A
1 Kit
Storage of components varies. See protocol for specific instructions.
Recommendations
Recommendations