Learn More
Novus Biologicals Human Adenylate Cyclase 3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP266450
Description
The Human Adenylate Cyclase 3 ELISA Kit (Colorimetric) measures human Adenylate Cyclase 3 in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes Human ADCY3 in samples. No significant cross-reactivity or interference between Human ADCY3 and analogues was observed. | |
109 | |
Adenylate Cyclase 3 | |
1 Kit | |
0.47 ng/mL |
0.78 - 50 ng/mL | |
ELISA | |
Storage is content dependent. | |
Human | |
Biological Fluids, Plasma, Serum | |
ADCY3 |