Learn More
Abnova™ Human ADIPOQ (NP_004788) Full-length Recombinant Protein
Used for SDS-PAGE
Manufacturer:  Abnova™ P4160
Catalog No. 89-932-705
Add to cart
Description
Description
This gene is expressed in adipose tissue exclusively. It encodes a protein with similarity to collagens X and VIII and complement factor C1q. The encoded protein circulates in the plasma and is involved with metabolic and hormonal processes. [provided by RefSeq]
Specifications
Specifications
NP_004788
Lyophilized
9370
ADIPOQ (Human) Recombinant Protein
25 ug
RUO
ACDC/ACRP30/ADPN/APM-1/APM1/GBP28/adiponectin
ADIPOQ
Yes
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized from 25mM Tris, 0.15 M NaCl, pH 7.4 (50 ug of BSA per 1μg of protein)
30kDa
≥90% by SDS-PAGE
Store at -20°C on dry atmosphere. After reconstitute with PBS containing at least 0.1% BSA, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
ADIPOQ
Escherichia coli
None