Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADPRHL1 Full-length ORF (NP_612439.2, 1 a.a. - 354 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00113622P01S

 View more versions of this product

Catalog No. 89-932-744


Add to cart

Description

Description

ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases (see ART1; MIM 601625) transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases, such as ADPRHL1, reverse the reaction (Glowacki et al., 2002 [PubMed 12070318]).[supplied by OMIM]

Sequence: MEKFKAAMLLGSVGDALGYRNVCKENSTVGMKIQEELQRSGGLDHLVLSPGEWPVSDNTIMHIATAEALTTDYWCLDDLYREMVRCYVEIVEKLPERRPDPATIEGCAQLKPNNYLLAWHTPFNEKGSGFGAATKAMCIGLRYWKPERLETLIEVSVECGRMTHNHPTGFLGSLCTALFVSFAAQGKPLVQWGRDMLRAVPLAEEYCRKTIRHTAEYQEHWFYFEAKWQFYLEERKISKDSENKAIFPDNYDAEEREKTYRKWSSEGRGGRRGHDAPMIAYDALLAAGNSWTELCHRAMFHGGESAATGTIAGCLFGLLYGLDLVPKGLYQDLEDKEKLEDLGAALYRLSTEEK
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ADPRHL1
Wheat Germ (in vitro)
GST
NP_612439.2
Liquid
113622
ADPRHL1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARH2
ADPRHL1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit