Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADSL Full-length ORF (NP_000017.1, 1 a.a. - 484 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000158P01S

 View more versions of this product

Catalog No. 89-932-763


Add to cart

Description

Description

Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAAGGDHGSPDSYRSPLASRYASPEMCFVFSDRYKFRTWRQLWLWLAEAEQTLGLPITDEQIQEMKSNLENIDFKMAAEEEKRLRHDVMAHVHTFGHCCPKAAGIIHLGATSCYVGDNTDLIILRNALDLLLPKLARVISRLADFAKERASLPTLGFTHFQPAQLTTVGKRCCLWIQDLCMDLQNLKRVRDDLRFRGVKGTTGTQASFLQLFEGDDHKVEQLDKMVTEKAGFKRAFIITGQTYTRKVDIEVLSVLASLGASVHKICTDIRLLANLKEMEEPFEKQQIGSSAMPYKRNPMRSERCCSLARHLMTLVMDPLQTASVQWFERTLDDSANRRICLAEAFLTADTILNTLQNISEGLVVYPKVIERRIRQELPFMATENIIMAMVKAGGSRQDCHEKIRVLSQQAASVVKQEGGDNDLIERIQVDAYFSPIHSQLDHLLDPSSFTGRASQQVQRFLEEEVYPLLKPYESVMKVKAELCL
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
81.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ADSL
Wheat Germ (in vitro)
GST
NP_000017.1
Liquid
158
ADSL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AMPS/ASASE/ASL
ADSL
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit