Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADSSL1 Full-length ORF (NP_689541.1, 1 a.a. - 457 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00122622P01L

 View more versions of this product

Catalog No. 89-932-773


Add to cart

Description

Description

ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP) (Sun et al., 2005 [PubMed 15786719]).[supplied by OMIM]

Sequence: MSGTRASNDRPPGAGGVKRGRLQQEAAATGSRVTVVLGAQWGDEGKGKVVDLLATDADIISRCQGGNNAGHTVVVDGKEYDFHLLPSGIINTKAVSFIGNGVVIHLPGLFEEAEKNEKKGLKDWEKRLIISDRAHLVFDFHQAVDGLQEVQRQAQEGKNIGTTKKGIGPTYSSKAARTGLRICDLLSDFDEFSSRFKNLAHQHQSMFPTLEIDIEGQLKRLKGFAERIRPMVRDGVYFMYEALHGPPKKILVEGANAALLDIDFGTYPFVTSSNCTVGGVCTGLGIPPQNIGDVYGVVKAYTTRVGIGAFPTEQINEIGGLLQTRGHEWGVTTGRKRRCGWLDLMILRYAHMVNGFTALALTKLDILDVLGEVKVGVSYKLNGKRIPYFPANQEMLQKVEVEYETLPGWKADTTGARRWEDLPPQAQNYIRFVENHVGVAVKWVGVGKSRESMIQLF
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ADSSL1
Wheat Germ (in vitro)
GST
NP_689541.1
Liquid
122622
ADSSL1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38602
ADSSL1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit