Learn More
Novus Biologicals™ Human AFAP ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266655
Catalog No. NBP266655
Add to Cart
Description
Description
The Human AFAP ELISA Kit (Colorimetric) measures human AFAP in serum, plasma and other biological fluids.
Specifications
Specifications
0.78 - 50 ng/mL
This kit recognizes Human AFAP1 in samples. No significant cross-reactivity or interference between Human AFAP1 and analogues was observed.
Biological Fluids, Plasma, Serum
60312
Human
Storage of components varies. See protocol for specific instructions.
0.47 ng/mL
ELISA Kit (Colorimetric)
ELISA
AFAP1
1 Kit
AFAP
Recommendations
Recommendations