Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human AGPAT7 Full-length ORF (NP_705841.2, 1 a.a. - 524 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00254531P01L
Description
Members of the 1-acylglycerol-3-phosphate O-acyltransferase (EC 2.3.1.51) family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction (Ye et al., 2005 [PubMed 16243729]).[supplied by OMIM]
Sequence: MSQGSPGDWAPLDPTPGPPASPNPFVHELHLSRLQRVKFCLLGALLAPIRVLLAFIVLFLLWPFAWLQVAGLSEEQLQEPITGWRKTVCHNGVLGLSRLLFFLLGFLRIRVRGQRASRLQAPVLVAAPHSTFFDPIVLLPCDLPKVVSRAENLSVPVIGALLRFNQAILVSRHDPASRRRVVEEVRRRATSGGKWPQVLFFPEGTCSNKKALLKFKPGAFIAGVPVQPVLIRYPNSLDTTSWAWRGPGVLKVLWLTASQPCSIVDVEFLPVYHPSPEESRDPTLYANNVQRVMAQALGIPATECEFVGSLPVIVVGRLKVALEPQLWELGKVLRKAGLSAGYVDAGAEPGRSRMISQEEFARQLQLSDPQTVAGAFGYFQQDTKGLVDFRDVALALAALDGGRSLEELTRLAFELFAEEQAEGPNRLLYKDGFSTILHLLLGSPHPAATALHAELCQAGSSQGLSLCQFQNFSLHDPLYGKLFSTYLRPPHTSRGTSQTPNASSPGNPTALANGTVQAPKQKGDSpecifications
NP_705841.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGPAT7/AYTL3/FLJ10257/LPAAT-eta/LPEAT2 | |
LPCAT4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
254531 | |
AGPAT7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQGSPGDWAPLDPTPGPPASPNPFVHELHLSRLQRVKFCLLGALLAPIRVLLAFIVLFLLWPFAWLQVAGLSEEQLQEPITGWRKTVCHNGVLGLSRLLFFLLGFLRIRVRGQRASRLQAPVLVAAPHSTFFDPIVLLPCDLPKVVSRAENLSVPVIGALLRFNQAILVSRHDPASRRRVVEEVRRRATSGGKWPQVLFFPEGTCSNKKALLKFKPGAFIAGVPVQPVLIRYPNSLDTTSWAWRGPGVLKVLWLTASQPCSIVDVEFLPVYHPSPEESRDPTLYANNVQRVMAQALGIPATECEFVGSLPVIVVGRLKVALEPQLWELGKVLRKAGLSAGYVDAGAEPGRSRMISQEEFARQLQLSDPQTVAGAFGYFQQDTKGLVDFRDVALALAALDGGRSLEELTRLAFELFAEEQAEGPNRLLYKDGFSTILHLLLGSPHPAATALHAELCQAGSSQGLSLCQFQNFSLHDPLYGKLFSTYLRPPHTSRGTSQTPNASSPGNPTALANGTVQAPKQKGD | |
RUO | |
LPCAT4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction