Learn More
Abnova™ Human AHI1 Full-length ORF (AAH94800.1, 1 a.a. - 609 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH94800.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54806 |
Molecular Weight (g/mol) | 96.4kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-932-898
|
Abnova™
H00054806P01S |
10 ug |
Each for $499.00
|
|
|||||
89-932-899
|
Abnova™
H00054806P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MPTAESEAKVKTKVRFEELLKTHSDLMREKKKLKKKLVRSEENISPDTIRSNLHYMKETTSDDPDTIRSNLPHIKETTSDDVSAANTNNLKKSTRVTKNKLRNTQLATENPNGDASVEEDKQGKPNKKVIKTVPQLTTQDLKPETPENKVDSTHQKTHTKPQPGVDHQKSEKANEGREETDLEEDEELMQAYQCHVTEEMAKEIKRKIRKKLKEQLTYFPSDTLFHDDKLSSEKRKKKKEVPVFSKAETSTLTISGDTVEGEQKKESSVRSVSSDSHQDDEISSMEQSTEDSMQDDTKPKPKKTKKKTKAVADNNEDVDGDGVHEITSRDSPVYPKCLLDDDLVLGVYIHRTDRLKSDFMISHPMVKIHVVDEHTGQYVKKDDSGRPVSSYYEKENVDYILPIMTQPYDFKQLKSRLPEWEEQIVFNENFPYLLRGSDESPKVILFFEILDFLSVDEIKNNSEVQNQECGFRKIAWAFLKLLGANGNANINSKLRLQLYYPPTKPRSPLSVVEAFEWWSKCPRNHYPSTLYVTVRGLKVPDCIKPSYRSMMALQEEKGKPVHCERHHESSSVDTEPGLEESKEVIKWKRLPGQENEDVFVLISPTMEEYSpecifications
AAH94800.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
96.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AHI-1/DKFZp686J1653/FLJ14023/FLJ20069/JBTS3/ORF1/dJ71N10.1 | |
AHI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54806 | |
AHI1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AHI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.