Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AHRR Partial ORF (NP_065782, 617 a.a. - 715 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name AHRR (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.63kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-932-917
View Documents
Abnova™
H00057491Q01S
10 ug
Each for $460.50
Add to cart
 
89-932-918
View Documents
Abnova™
H00057491Q01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

Dioxin is a teratogen that exerts its effects through the arylhydrocarbon receptor in conjunction with the receptor's binding partner, arylhydrocarbon receptor nuclear translocator. The protein encoded by this gene represses signal transduction by the arylhydrocarbon receptor by competing with the arylhydrocarbon receptor nuclear translocator for binding to the arylhydrocarbon receptor. Expression of the repressor is stimulated by the receptor/translocator heterodimer, thereby regulating receptor function through a negative feedback mechanism. In addition, the encoded protein can bind to nuclear factor kappa-B. [provided by RefSeq]

Sequence: RATAGRSRELTPFHPAHCACLEPTDGLPQSEPPHQLCARGRGEQSCTCRAAEAAPVVKREPLDSPQWATHSQGMVPGMLPKSALATLVPPQASGCTFLP
Specifications

Specifications

AHRR (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.63kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit