Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aiolos/IKZF3 Alexa Fluor 594-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8625T100UG
This item is not returnable.
View return policy
Description
Aiolos/IKZF3 Monoclonal specifically detects Aiolos/IKZF3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Aiolos/IKZF3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UKT9 | |
IKZF3 | |
E. coli-derived recombinant human Aiolos/IKZF3 Met1-Gly54 Accession # Q9UKT9 | |
100 μg | |
Primary | |
Detects human Aiolos/IKZF3 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
914821 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, IKZF3, zinc finger protein Aiolos, ZNFN1A3, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
22806 | |
Human | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction