Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AK3L1 Full-length ORF (NP_001002921.1, 1 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000205P02S

 View more versions of this product

Catalog No. 89-932-953


Add to cart

Description

Description

This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1. [provided by RefSeq]

Sequence: MASKLLRAVILGPPGSGKGTVCQRIAQNFGLQHLSSGHFLRENIKASTEVGEMAKQYIEKSLLVPDHVITRLMMSELENRRGQHWLLDGFPRTLGQAEALDKICEVDLVISLNIPFETLKDRLSRRWIHPPSGRVYNLDFNPPHVHGIDDVTGEPLVQQEDDKPEAVAARLRQYKDVAKPVIELYKSRGVLHQFSGTETNKIWPYVYTLFSNKITPIQSKEAY
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AK3L1
Wheat Germ (in vitro)
GST
NP_001002921.1
Liquid
205
AK3L1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AK3/AK4/MGC166959
AK3L1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit