Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKAP1 Partial ORF (NP_003479, 806 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008165Q01S

 View more versions of this product

Catalog No. 89-932-962


Add to cart

Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein binds to type I and type II regulatory subunits of PKA and anchors them to the mitochondrion. This protein is speculated to be involved in the cAMP-dependent signal transduction pathway and in directing RNA to a specific cellular compartment. [provided by RefSeq]

Sequence: VLRQIRSDFVTLPFQGAEVLLDSVMPLSDDDQFSPEADAAMSEMTGNTALLAQVTSYSPTGLPLIQLWSVVGDEVVLINRSLVERGLAQWVDSYYTSL
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VLRQIRSDFVTLPFQGAEVLLDSVMPLSDDDQFSPEADAAMSEMTGNTALLAQVTSYSPTGLPLIQLWSVVGDEVVLINRSLVERGLAQWVDSYYTSL
RUO
AKAP1
Wheat Germ (in vitro)
GST
NP_003479
Liquid
8165
AKAP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKAP/AKAP121/AKAP149/AKAP84/D-AKAP1/MGC1807/PRKA1/SAKAP84
AKAP1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit