Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKAP10 Full-length ORF (NP_009133.2, 1 a.a. - 662 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011216P01L

 View more versions of this product

Catalog No. 89-932-968


Add to cart

Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS (regulator of G protein signalling) domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction. [provided by RefSeq]

Sequence: MRGAGPSPRQSPRTLRPDPGPAMSFFRRKVKGKEQEKTSDVKSIKASISVHSPQKSTKNHALLEAAGPSHVAINAISANMDSFSSSRTATLKKQPSHMEAAHFGDLGRSCLDYQTQETKSSLSKTLEQVLHDTIVLPYFIQFMELRRMEHLVKFWLEAESFHSTTWSRIRAHSLNTVKQSSLAEPVSPSKKHETTASFLTDSLDKRLEDSGSAQLFMTHSEGIDLNNRTNSTQNHLLLSQECDSAHSLRLEMARAGTHQVSMETQESSSTLTVASRNSPASPLKELSGKLMKSIEQDAVNTFTKYISPDAAKPIPITEAMRNDIIARICGEDGQVDPNCFVLAQSIVFSAMEQEHFSEFLRSHHFCKYQIEVLTSGTVYLADILFCESALFYFSEYMEKEDAVNILQFWLAADNFQSQLAAKKGQYDGQEAQNDAMILYDKYFSLQATHPLGFDDVVRLEIESNICREGGPLPNCFTTPLRQAWTTMEKVFLPGFLSSNLYYKYLNDLIHSVRGDEFLGGNVSLTAPGSVGPPDESHPGSSDSSASQSSVKKASIKILKNFDEAIIVDAASLDPESLYQRTYAGKMTFGRVSDLGQFIRESEPEPDVRKSKGSMFSQAMKKWVQGNTDEAQEELAWKIAKMIVSDIMQQAQYDQPLEKSTKL
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
100.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AKAP10
Wheat Germ (in vitro)
GST
NP_009133.2
Liquid
11216
AKAP10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D-AKAP2/MGC9414/PRKA10
AKAP10
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit