Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKAP11 Partial ORF (NP_057332, 1801 a.a. - 1901 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011215Q01S

 View more versions of this product

Catalog No. 89-932-971


Add to cart

Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. [provided by RefSeq]

Sequence: EGLGQDGKTLLITNIDMEPCTVDPQLRIILQWLIASEAEVAELYFHDSANKEFMLLSKQLQEKGWKVGDLLQAVLQYYEVMEKASSEERCKSLFDWLLENA
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EGLGQDGKTLLITNIDMEPCTVDPQLRIILQWLIASEAEVAELYFHDSANKEFMLLSKQLQEKGWKVGDLLQAVLQYYEVMEKASSEERCKSLFDWLLENA
RUO
AKAP11
Wheat Germ (in vitro)
GST
NP_057332
Liquid
11215
AKAP11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKAP220/DKFZp781I12161/FLJ11304/KIAA0629/PRKA11
AKAP11
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit