Learn More
Abnova™ Human AKAP14 Full-length ORF (NP_848928.1, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00158798P01S
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The protein anchors PKA in ciliary axonemes and, in this way, may play a role in regulating ciliary beat frequency. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MSETQNSTSQKAMDEDNKAASQTMPNTQDKNYEDELTQVALALVEDVINYAVKIVEEERNPLKNIKWMTHGEFTVEKGLKQIDEYFSKCVSKKCWAHGVEFVERKDLIHSFLYIYYVHWSISTADLPVARISAGTYFTMKVSKTKPPDAPIVVSYVGDHQALVHRPGMVRFRENWQKNLTDAKYSFMESFPFLFNRVSpecifications
NP_848928.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AKAP14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
158798 | |
AKAP14 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKAP28 | |
AKAP14 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.