Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKAP6 Partial ORF (NP_004265.3, 2221 a.a. - 2318 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009472Q01S

 View more versions of this product

Catalog No. 89-932-987


Add to cart

Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is highly expressed in various brain regions and cardiac and skeletal muscle. It is specifically localized to the sarcoplasmic reticulum and nuclear membrane, and is involved in anchoring PKA to the nuclear membrane or sarcoplasmic reticulum. [provided by RefSeq]

Sequence: RAEVGKEVNGLPQTSSGCAENLEFTPSKLDSEKESSGKPGESGMPEEHNAASAKSKVQDLSLKANQPTDKAALHPSPKTLTCEENLLNLHEKRHRNMH
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RAEVGKEVNGLPQTSSGCAENLEFTPSKLDSEKESSGKPGESGMPEEHNAASAKSKVQDLSLKANQPTDKAALHPSPKTLTCEENLLNLHEKRHRNMH
RUO
AKAP6
Wheat Germ (in vitro)
GST
NP_004265.3
Liquid
9472
AKAP6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADAP100/ADAP6/AKAP100/KIAA0311/MGC165020/PRKA6/mAKAP
AKAP6
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit